Anti-DNAAF1 Mouse Monoclonal Antibody [clone: OTI3A1]

Supplier: OriGene
ORIGTA504575EA 603 EUR
ORIGTA504575
Anti-DNAAF1 Mouse Monoclonal Antibody [clone: OTI3A1]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DNAAF1 (dynein, axonemal, assembly factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR