Anti-ATRIP Mouse Monoclonal Antibody [clone: OTI1F10]

Supplier: OriGene
TA504586
ORIGTA504586EA 616 EUR
ORIGTA504586
Anti-ATRIP Mouse Monoclonal Antibody [clone: OTI1F10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ATRIP (ATR interacting protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR