Anti-SPINT1 Mouse Monoclonal Antibody [clone: OTI4H2]

Supplier: OriGene
ORIGTA504591EA 621 EUR
ORIGTA504591
Anti-SPINT1 Mouse Monoclonal Antibody [clone: OTI4H2]
Antibodies
Recommended Dilutions: Western Blot: 1:200; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SPINT1 (serine peptidase inhibitor, Kunitz type 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR