Anti-CRYZL1 Mouse Monoclonal Antibody [clone: OTI1A3]

Supplier: OriGene
ORIGTA504599EA 603 EUR
ORIGTA504599
Anti-CRYZL1 Mouse Monoclonal Antibody [clone: OTI1A3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CRYZL1 (crystallin, zeta (quinone reductase)-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR