Anti-RBM46 Mouse Monoclonal Antibody [clone: OTI1D6]

Supplier: OriGene
ORIGTA504623EA 586 EUR
ORIGTA504623
Anti-RBM46 Mouse Monoclonal Antibody [clone: OTI1D6]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RBM46 (RNA binding motif protein 46)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR