Anti-ATRIP Mouse Monoclonal Antibody [clone: OTI4A8]

Supplier: OriGene
ORIGTA504640EA 603 EUR
ORIGTA504640
Anti-ATRIP Mouse Monoclonal Antibody [clone: OTI4A8]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: ATRIP (ATR interacting protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR