Anti-SCFD1 Mouse Monoclonal Antibody [clone: OTI3F2]

Supplier: OriGene
ORIGTA504693EA 603 EUR
ORIGTA504693
Anti-SCFD1 Mouse Monoclonal Antibody [clone: OTI3F2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: SCFD1 (sec1 family domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR