Anti-METTL16 Mouse Monoclonal Antibody [clone: OTI5F10]

Supplier: OriGene
ORIGTA504697EA 581 EUR
ORIGTA504697
Anti-METTL16 Mouse Monoclonal Antibody [clone: OTI5F10]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: METTL16 (methyltransferase like 16)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR