Anti-MLF1 Mouse Monoclonal Antibody [clone: OTI2D6]

Supplier: OriGene
ORIGTA504765EA 603 EUR
ORIGTA504765
Anti-MLF1 Mouse Monoclonal Antibody [clone: OTI2D6]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MLF1 (myeloid leukemia factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR