Anti-MPI Mouse Monoclonal Antibody [clone: OTI4A11]

Supplier: OriGene
ORIGTA504771EA 603 EUR
ORIGTA504771
Anti-MPI Mouse Monoclonal Antibody [clone: OTI4A11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MPI (mannose phosphate isomerase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR