Anti-NQO2 Mouse Monoclonal Antibody [clone: OTI5F6]

Supplier: OriGene
ORIGTA504782EA 603 EUR
ORIGTA504782
Anti-NQO2 Mouse Monoclonal Antibody [clone: OTI5F6]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: NQO2 (NAD(P)H dehydrogenase, quinone 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR