Anti-DOK7 Mouse Monoclonal Antibody [clone: OTI1F7]

Supplier: OriGene
TA504789
ORIGTA504789EA 616 EUR
ORIGTA504789
Anti-DOK7 Mouse Monoclonal Antibody [clone: OTI1F7]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DOK7 (docking protein 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR