Anti-PECAM1 Mouse Monoclonal Antibody [clone: OTI3B5]

Supplier: OriGene
ORIGTA504803EA 616 EUR
ORIGTA504803
Anti-PECAM1 Mouse Monoclonal Antibody [clone: OTI3B5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR