Anti-ACBD3 Mouse Monoclonal Antibody [clone: OTI1G2]

Supplier: OriGene
ORIGTA504822EA 616 EUR
ORIGTA504822
Anti-ACBD3 Mouse Monoclonal Antibody [clone: OTI1G2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ACBD3 (acyl-CoA binding domain containing 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR