Anti-MLF1 Mouse Monoclonal Antibody [clone: OTI5E11]

Supplier: OriGene
TA504826
ORIGTA504826EA 616 EUR
ORIGTA504826
Anti-MLF1 Mouse Monoclonal Antibody [clone: OTI5E11]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MLF1 (myeloid leukemia factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR