Anti-MLF2 Mouse Monoclonal Antibody [clone: OTI1G1]

Supplier: OriGene
ORIGTA504876EA 581 EUR
ORIGTA504876
Anti-MLF2 Mouse Monoclonal Antibody [clone: OTI1G1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MLF2 (myeloid leukemia factor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR