Anti-COMMD1 Mouse Monoclonal Antibody [clone: OTI3B3]

Supplier: OriGene
ORIGTA504889EA 603 EUR
ORIGTA504889
Anti-COMMD1 Mouse Monoclonal Antibody [clone: OTI3B3]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: COMMD1 (copper metabolism (Murr1) domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR