Anti-PADI4 Mouse Monoclonal Antibody [clone: OTI5B7]

Supplier: OriGene
ORIGTA504897EA 603 EUR
ORIGTA504897
Anti-PADI4 Mouse Monoclonal Antibody [clone: OTI5B7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PADI4 (peptidyl arginine deiminase, type IV)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR