Anti-SNAI2 Mouse Monoclonal Antibody [clone: OTI2B12]

Supplier: OriGene
ORIGTA504907EA 603 EUR
ORIGTA504907
Anti-SNAI2 Mouse Monoclonal Antibody [clone: OTI2B12]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: SNAI2 (snail family zinc finger 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR