Anti-VBP1 Mouse Monoclonal Antibody [clone: OTI2E6]

Supplier: OriGene
ORIGTA504914EA 603 EUR
ORIGTA504914
Anti-VBP1 Mouse Monoclonal Antibody [clone: OTI2E6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: VBP1 (von Hippel-Lindau binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR