Anti-FTCD Mouse Monoclonal Antibody [clone: OTI1A7]

Supplier: OriGene
ORIGTA504944EA 581 EUR
ORIGTA504944
Anti-FTCD Mouse Monoclonal Antibody [clone: OTI1A7]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: FTCD (formimidoyltransferase cyclodeaminase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR