Anti-APP Mouse Monoclonal Antibody [clone: OTI1E6]

Supplier: OriGene
TA504968
ORIGTA504968EA 616 EUR
ORIGTA504968
Anti-APP Mouse Monoclonal Antibody [clone: OTI1E6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR