Anti-POFUT2 Mouse Monoclonal Antibody [clone: OTI3A12]

Supplier: OriGene
ORIGTA504972EA 603 EUR
ORIGTA504972
Anti-POFUT2 Mouse Monoclonal Antibody [clone: OTI3A12]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: POFUT2 (protein O-fucosyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR