Anti-UBE2J1 Mouse Monoclonal Antibody [clone: OTI1H10]

Supplier: OriGene
ORIGTA504974EA 603 EUR
ORIGTA504974
Anti-UBE2J1 Mouse Monoclonal Antibody [clone: OTI1H10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: UBE2J1 (ubiquitin-conjugating enzyme E2, J1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR