Anti-UBE2J1 Mouse Monoclonal Antibody [clone: OTI3D5]

Supplier: OriGene
ORIGTA505006EA 603 EUR
ORIGTA505006
Anti-UBE2J1 Mouse Monoclonal Antibody [clone: OTI3D5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: UBE2J1 (ubiquitin-conjugating enzyme E2, J1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR