Anti-TMEM173 Mouse Monoclonal Antibody [clone: OTI2C8]

Supplier: OriGene
ORIGTA505020EA 603 EUR
ORIGTA505020
Anti-TMEM173 Mouse Monoclonal Antibody [clone: OTI2C8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TMEM173 (transmembrane protein 173)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR