Anti-RFC2 Mouse Monoclonal Antibody [clone: OTI2B2]

Supplier: OriGene
ORIGTA505033EA 581 EUR
ORIGTA505033
Anti-RFC2 Mouse Monoclonal Antibody [clone: OTI2B2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: RFC2 (replication factor C (activator 1) 2, 40kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR