Anti-ZBED1 Mouse Monoclonal Antibody [clone: OTI2A7]

Supplier: OriGene
TA505040
ORIGTA505040EA 616 EUR
ORIGTA505040
Anti-ZBED1 Mouse Monoclonal Antibody [clone: OTI2A7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ZBED1 (zinc finger, BED-type containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR