Anti-GORAB Mouse Monoclonal Antibody [clone: OTI4F11]

Supplier: OriGene
ORIGTA505074EA 603 EUR
ORIGTA505074
Anti-GORAB Mouse Monoclonal Antibody [clone: OTI4F11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: GORAB (golgin, RAB6-interacting)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR