Anti-GORAB Mouse Monoclonal Antibody [clone: OTI1E3]

Supplier: OriGene
ORIGTA505075EA 608 EUR
ORIGTA505075
Anti-GORAB Mouse Monoclonal Antibody [clone: OTI1E3]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: GORAB (golgin, RAB6-interacting)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR