Anti-ESRRG Mouse Monoclonal Antibody [clone: OTI1E5]

Supplier: OriGene
ORIGTA505080EA 608 EUR
ORIGTA505080
Anti-ESRRG Mouse Monoclonal Antibody [clone: OTI1E5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: ESRRG (estrogen-related receptor gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR