Anti-CLEC4E Mouse Monoclonal Antibody [clone: OTI2H4]

Supplier: OriGene
ORIGTA505097EA 603 EUR
ORIGTA505097
Anti-CLEC4E Mouse Monoclonal Antibody [clone: OTI2H4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CLEC4E (C-type lectin domain family 4, member E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR