Anti-CHN1 Mouse Monoclonal Antibody [clone: OTI2D10]

Supplier: OriGene
TA505103
ORIGTA505103EA 616 EUR
ORIGTA505103
Anti-CHN1 Mouse Monoclonal Antibody [clone: OTI2D10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CHN1 (chimerin 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR