Anti-PROSC Mouse Monoclonal Antibody [clone: OTI3H4]

Supplier: OriGene
ORIGTA505130EA 581 EUR
ORIGTA505130
Anti-PROSC Mouse Monoclonal Antibody [clone: OTI3H4]
Antibodies
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: PROSC (proline synthetase co-transcribed homolog (bacterial))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR