Anti-RIOK2 Mouse Monoclonal Antibody [clone: OTI3E11]

Supplier: OriGene
TA505140
ORIGTA505140EA 598 EUR
ORIGTA505140
Anti-RIOK2 Mouse Monoclonal Antibody [clone: OTI3E11]
Antibodies
Recommended Dilutions: Western Blot: 1:500-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RIOK2 (RIO kinase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR