Anti-OTUB1 Mouse Monoclonal Antibody [clone: OTI1E3]

Supplier: OriGene
TA505157
ORIGTA505157EA 616 EUR
ORIGTA505157
Anti-OTUB1 Mouse Monoclonal Antibody [clone: OTI1E3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: OTUB1 (OTU deubiquitinase, ubiquitin aldehyde binding 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR