Anti-STK38L Mouse Monoclonal Antibody [clone: OTI4D8]

Supplier: OriGene
TA505176
ORIGTA505176EA 641 EUR
ORIGTA505176
Anti-STK38L Mouse Monoclonal Antibody [clone: OTI4D8]
Antibodies
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: STK38L (serine/threonine kinase 38 like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR