Anti-FATE1 Mouse Monoclonal Antibody [clone: OTI3D7]

Supplier: OriGene
ORIGTA505182EA 603 EUR
ORIGTA505182
Anti-FATE1 Mouse Monoclonal Antibody [clone: OTI3D7]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FATE1 (fetal and adult testis expressed 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR