Anti-ZC2HC1A Mouse Monoclonal Antibody [clone: OTI4C1]

Supplier: OriGene
TA505191
ORIGTA505191EA 621 EUR
ORIGTA505191
Anti-ZC2HC1A Mouse Monoclonal Antibody [clone: OTI4C1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ZC2HC1A (zinc finger, C2HC-type containing 1A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR