Anti-CDKN2B Mouse Monoclonal Antibody [clone: OTI7C4]

Supplier: OriGene
ORIGTA505205EA 603 EUR
ORIGTA505205
Anti-CDKN2B Mouse Monoclonal Antibody [clone: OTI7C4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CDKN2B (cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR