Anti-MRPS34 Mouse Monoclonal Antibody [clone: OTI4B9]

Supplier: OriGene
TA505233
ORIGTA505233EA 616 EUR
ORIGTA505233
Anti-MRPS34 Mouse Monoclonal Antibody [clone: OTI4B9]
Antibodies
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MRPS34 (mitochondrial ribosomal protein S34)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR