Anti-CCT8L2 Mouse Monoclonal Antibody [clone: OTI1A8]

Supplier: OriGene
ORIGTA505336EA 593 EUR
ORIGTA505336
Anti-CCT8L2 Mouse Monoclonal Antibody [clone: OTI1A8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CCT8L2 (chaperonin containing TCP1, subunit 8 (theta)-like 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR