Anti-TRMT2A Mouse Monoclonal Antibody [clone: OTI1G8]

Supplier: OriGene
TA505516
ORIGTA505516EA 593 EUR
ORIGTA505516
Anti-TRMT2A Mouse Monoclonal Antibody [clone: OTI1G8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TRMT2A (tRNA methyltransferase 2 homolog A (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR