Anti-GADD45G Mouse Monoclonal Antibody [clone: OTI4E5]

Supplier: OriGene
TA505533
ORIGTA505533EA 616 EUR
ORIGTA505533
Anti-GADD45G Mouse Monoclonal Antibody [clone: OTI4E5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GADD45G (growth arrest and DNA-damage-inducible, gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR