Anti-GLB1 Mouse Monoclonal Antibody [clone: OTI2F6]

Supplier: OriGene
TA505568
ORIGTA505568EA 616 EUR
ORIGTA505568
Anti-GLB1 Mouse Monoclonal Antibody [clone: OTI2F6]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: GLB1 (galactosidase, beta 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR