Anti-NDOR1 Mouse Monoclonal Antibody [clone: OTI2G4]

Supplier: OriGene
TA505579
ORIGTA505579EA 616 EUR
ORIGTA505579
Anti-NDOR1 Mouse Monoclonal Antibody [clone: OTI2G4]
Antibodies
Recommended Dilutions: Western Blot: 1:200-1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NDOR1 (NADPH dependent diflavin oxidoreductase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR