Anti-MAP2K1 Mouse Monoclonal Antibody [clone: OTI7F3]

Supplier: OriGene
TA506032
ORIGTA506032EA 616 EUR
ORIGTA506032
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: OTI7F3]
Antibodies
Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR