Anti-MSI1 Mouse Monoclonal Antibody [clone: OTI3C2]

Supplier: OriGene
TA506364
ORIGTA506364EA 616 EUR
ORIGTA506364
Anti-MSI1 Mouse Monoclonal Antibody [clone: OTI3C2]
Antibodies
  • Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: MSI1 (musashi RNA-binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR