Anti-TH Mouse Monoclonal Antibody [clone: OTI3A7]

Supplier: OriGene
TA506548
ORIGTA506548EA 616 EUR
ORIGTA506548
Anti-TH Mouse Monoclonal Antibody [clone: OTI3A7]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR