Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI1G4]

Supplier: OriGene
ORIGTA506657EA 616 EUR
ORIGTA506657
Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI1G4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR